The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018340.2 | NP_060810.2 | 3 - 4 | 66 | |
GenBank mRNA | AK022710.1 | - | 3 - 4 | 774 | 66 |
AK093854.1 | - | 3 - 4 | 570 | 66 | |
AK225359.1 | - | 3 - 4 | 774 | 66 | |
AK225617.1 | - | 3 - 4 | 819 | 66 | |
AY553877.1 | - | 3 - 4 | 713 | 66 | |
BC006289.2 | - | 3 - 4 | 808 | 66 |