The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135155.2 | NP_001128627.1 | 5 - 6 | 625 | 93 |
NM_001135156.2 | NP_001128628.1 | 5 - 6 | 639 | 93 | |
NM_001289978.1 | NP_001276907.1 | 5 - 6 | 625 | 93 | |
NM_004647.3 | NP_004638.2 | 5 - 6 | 625 | 93 | |
XM_005259289.2 | XP_005259346.1 | 5 - 6 | 625 | 93 | |
XM_005259290.3 | XP_005259347.1 | 5 - 6 | 596 | 93 | |
XM_005259292.4 | XP_005259349.1 | 5 - 6 | 562 | 93 | |
XM_006723407.2 | XP_006723470.1 | 5 - 6 | 596 | 93 | |
XM_006723408.3 | XP_006723471.1 | 5 - 6 | 564 | 93 | |
XM_006723409.2 | XP_006723472.1 | 5 - 6 | 625 | 93 | |
XM_006723410.2 | XP_006723473.1 | 2 - 3 | 177 | 93 | |
XM_011527356.1 | XP_011525658.1 | 5 - 6 | 625 | 93 | |
XM_011527357.2 | XP_011525659.1 | 5 - 6 | 565 | 93 | |
XM_011527358.2 | XP_011525660.1 | 5 - 6 | 640 | 93 | |
GenBank mRNA | AK094632.1 | - | 5 - 6 | 952 | 93 |
AK127198.1 | - | 2 - 3 | 1099 | 93 | |
AK297581.1 | - | 6 - 7 | 599 | 93 | |
AK308563.1 | - | 5 - 6 | 625 | 93 | |
AL529673.3 | - | 5 - 6 | 606 | 92 | |
AL538568.3 | - | 5 - 6 | 644 | 93 | |
BC021191.2 | - | 5 - 6 | 598 | 93 | |
BC125152.1 | - | 5 - 6 | 527 | 93 | |
BC125153.1 | - | 5 - 6 | 527 | 93 | |
GQ358728.1 | - | 5 - 6 | 517 | 93 | |
U43843.1 | - | 5 - 6 | 523 | 93 |