The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004647.3 | NP_004638.2 | 6 - 7 | 699 | 62 |
GenBank mRNA | U43843.1 | - | 6 - 7 | 597 | 62 |