Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAHD1
Entrez Gene ID
22893
Gene Name
bromo adjacent homology domain containing 1
Gene Aliases
-
Chromosome Location
Chr.15: 40439721 - 40468242 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301132.1 NP_001288061.1 4 - 5 62
NM_014952.4 NP_055767.3 4 - 5 62
NR_125372.1 2 - 3 62
XM_011521366.2 XP_011519668.2 4 - 5 62
XM_011521367.2 XP_011519669.2 4 - 5 62
XM_011521368.2 XP_011519670.2 4 - 5 62
XM_011521369.2 XP_011519671.1 4 - 5 62
GenBank mRNA AB023162.1 - 4 - 5 62
AL833923.1 - 3 - 4 62
BC022782.2 - 4 - 5 62
HY353002.1 - 3 - 4 262 62

View Full Product Details