The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002069.5 | NP_002060.4 | 2 - 3 | 496 | 107 |
GenBank mRNA | AF055013.1 | - | 2 - 3 | 171 | 107 |
AF493905.1 | - | 2 - 3 | 159 | 107 | |
AK292953.1 | - | 2 - 3 | 470 | 107 | |
AL049933.1 | - | 2 - 3 | 474 | 107 | |
BC026326.1 | - | 2 - 3 | 495 | 107 | |
BT019775.1 | - | 2 - 3 | 159 | 107 | |
M17219.1 | - | 2 - 3 | 144 | 107 |