Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR97
Entrez Gene ID
340390
Gene Name
WD repeat domain 97
Gene Aliases
KIAA1875
Chromosome Location
Chr.8: 144107692 - 144116392 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316309.1 NP_001303238.1 20 - 21 3976 66
XM_017013385.1 XP_016868874.1 19 - 20 4099 66
XM_017013386.1 XP_016868875.1 19 - 20 4099 66
XM_017013387.1 XP_016868876.1 19 - 20 4040 66
XM_017013388.1 XP_016868877.1 19 - 20 4048 66
XM_017013389.1 XP_016868878.1 20 - 21 4021 66
XM_017013390.1 XP_016868879.1 19 - 20 4003 66
XM_017013391.1 XP_016868880.1 18 - 19 3973 66
XM_017013392.1 XP_016868881.1 18 - 19 3970 66
XM_017013393.1 XP_016868882.1 19 - 20 3937 66
XM_017013394.1 XP_016868883.1 18 - 19 3885 66
GenBank mRNA AB058778.1 - 19 - 20 4145 66
AK095597.1 - 8 - 9 3262 66
AK124611.1 - 6 - 7 3452 66
AL137446.1 - 3 - 4 2038 66


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DR006545.1 - 1 - 2 453 Chr. 8 - 144114229 - 144115448 [+] on Build GRCh38 66


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream MAF1 homolog, negative regulator of RNA polymerase III MAF1 -6.9kb Sense Chr. - 144104279 - 144107612 on Build GRCh38
Downstream HGH1 homolog HGH1 BRP16, BRP16L, C8orf30A, C8orf30B, FAM203A, FAM203B 22.9kb Sense Chr. - 144137721 - 144140843 on Build GRCh38

View Full Product Details