The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001254755.1 | NP_001241684.1 | 3 - 4 | 472 | 100 |
NM_001254756.1 | NP_001241685.1 | 3 - 4 | 475 | 100 | |
NM_144611.3 | NP_653212.1 | 3 - 4 | 1167 | 100 | |
XM_006721447.2 | XP_006721510.2 | 3 - 4 | 2304 | 100 | |
XM_011523649.2 | XP_011521951.1 | 3 - 4 | 555 | 100 | |
XM_017024180.1 | XP_016879669.1 | 3 - 4 | 558 | 100 | |
XM_017024181.1 | XP_016879670.1 | 3 - 4 | 447 | 100 | |
XM_017024182.1 | XP_016879671.1 | 3 - 4 | 2609 | 100 | |
GenBank mRNA | AA479992.1 | - | 2 - 3 | 274 | 99 |
AK172844.1 | - | 3 - 4 | 633 | 100 | |
AK313088.1 | - | 3 - 4 | 638 | 100 | |
BC020263.1 | - | 3 - 4 | 619 | 100 | |
BC051697.1 | - | 3 - 4 | 1079 | 100 | |
BX327481.2 | - | 2 - 3 | 258 | 100 | |
CB112805.1 | - | 3 - 4 | 475 | 100 | |
CB129650.1 | - | 3 - 4 | 442 | 100 | |
CK821986.1 | - | 2 - 3 | 339 | 100 |