Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIGB
Entrez Gene ID
9488
Gene Name
phosphatidylinositol glycan anchor biosynthesis class B
Gene Aliases
GPI-MT-III, PIG-B
Chromosome Location
Chr.15: 55318935 - 55355648 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004855.4 NP_004846.4 2 - 3 613 71
XM_011522235.2 XP_011520537.1 2 - 3 613 71
XM_011522236.2 XP_011520538.1 2 - 3 613 71
GenBank mRNA AK223330.1 - 2 - 3 318 71
AK314706.1 - 2 - 3 588 71
BC017711.1 - 2 - 3 550 71
D42138.1 - 2 - 3 341 71

View Full Product Details