The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004855.4 | NP_004846.4 | 5 - 6 | 972 | 128 |
XM_005254795.4 | XP_005254852.1 | 3 - 4 | 401 | 128 | |
XM_011522235.2 | XP_011520537.1 | 5 - 6 | 972 | 128 | |
XM_011522236.2 | XP_011520538.1 | 4 - 5 | 867 | 128 | |
XM_011522237.2 | XP_011520539.1 | 4 - 5 | 476 | 128 | |
XM_017022730.1 | XP_016878219.1 | 4 - 5 | 391 | 128 | |
XM_017022731.1 | XP_016878220.1 | 5 - 6 | 520 | 128 | |
XM_017022732.1 | XP_016878221.1 | 3 - 4 | 401 | 128 | |
GenBank mRNA | AK293666.1 | - | 2 - 3 | 588 | 128 |
AK314706.1 | - | 5 - 6 | 947 | 128 | |
BC017711.1 | - | 5 - 6 | 909 | 128 | |
D42138.1 | - | 5 - 6 | 700 | 128 |