Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGAP1
Entrez Gene ID
392
Gene Name
Rho GTPase activating protein 1
Gene Aliases
CDC42GAP, RHOGAP, RHOGAP1, p50rhoGAP
Chromosome Location
Chr.11: 46677075 - 46700665 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004308.3 NP_004299.1 2 - 3 295 81
XM_011520095.1 XP_011518397.1 2 - 3 732 81
XM_017017720.1 XP_016873209.1 1 - 2 180 81
GenBank mRNA AK126404.1 - 2 - 3 732 81
AK298562.1 - 2 - 3 242 81
AK307296.1 - 2 - 3 233 81
AK310605.1 - 2 - 3 245 81
BC018118.1 - 2 - 3 200 81
HY004921.1 - 2 - 3 296 81
U02570.1 - 2 - 3 202 81
Z23024.1 - 2 - 3 229 81

View Full Product Details