Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POMT1
Entrez Gene ID
10585
Gene Name
protein O-mannosyltransferase 1
Gene Aliases
LGMD2K, MDDGA1, MDDGB1, MDDGC1, RT
Chromosome Location
Chr.9: 131502895 - 131523806 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077365.1 NP_001070833.1 1 - 2 173 95
NM_007171.3 NP_009102.3 1 - 2 173 95
XM_005272162.2 XP_005272219.1 1 - 2 147 95
XM_006716932.1 XP_006716995.1 1 - 2 58 95
XM_011518140.1 XP_011516442.1 1 - 2 173 95
XM_011518141.1 XP_011516443.1 1 - 2 173 95
XM_011518143.1 XP_011516445.1 1 - 2 173 95
GenBank mRNA AF095136.1 - 1 - 2 150 95
AK074874.1 - 1 - 2 131 95
AK300393.1 - 1 - 2 131 95
AK301486.1 - 1 - 2 155 95
AK303254.1 - 1 - 2 131 95
AK307876.1 - 1 - 2 131 95
AK315218.1 - 1 - 2 108 95
BC022877.2 - 1 - 2 67 95
DB154785.1 - 1 - 2 173 95

View Full Product Details