Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- POMT1
- Entrez Gene ID
-
10585
- Gene Name
- protein O-mannosyltransferase 1
- Gene Aliases
-
LGMD2K, MDDGA1, MDDGB1, MDDGC1, RT
- Chromosome Location
-
Chr.9: 131502895 - 131523806 on Build GRCh38
View Full Product Details