Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VARS2
Entrez Gene ID
57176
Gene Name
valyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD20, VALRS, VARS2L, VARSL
Chromosome Location
Chr.6: 30914208 - 30926459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167733.2 NP_001161205.1 19 - 20 1870 87
NM_001167734.1 NP_001161206.1 20 - 21 2105 87
NM_020442.5 NP_065175.4 20 - 21 2098 87
GenBank mRNA AB067472.1 - 19 - 20 2566 87
AK000511.1 - 19 - 20 1775 87
AK025618.1 - 8 - 9 690 87
AK293780.1 - 19 - 20 1859 87
AK303415.1 - 20 - 21 2087 87
AK304807.1 - 20 - 21 2105 87
AK315733.1 - 9 - 10 856 87
BC008844.2 - 3 - 4 293 87
BC009355.2 - 10 - 11 946 87
BC063427.1 - 8 - 9 721 87
BC073838.1 - 8 - 9 716 87
BC112054.1 - 19 - 20 1960 87
BC113605.1 - 19 - 20 1960 87
BC143534.1 - 18 - 19 1888 87
BC143535.1 - 19 - 20 1959 87
BC143536.1 - 19 - 20 1953 87

View Full Product Details