The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167733.2 | NP_001161205.1 | 23 - 24 | 2248 | 102 |
NM_001167734.1 | NP_001161206.1 | 24 - 25 | 2483 | 102 | |
NM_020442.5 | NP_065175.4 | 24 - 25 | 2476 | 102 | |
GenBank mRNA | AB067472.1 | - | 23 - 24 | 2944 | 102 |
AK000511.1 | - | 23 - 24 | 2153 | 102 | |
AK025618.1 | - | 12 - 13 | 1068 | 102 | |
AK026063.1 | - | 3 - 4 | 256 | 102 | |
AK125069.1 | - | 13 - 14 | 2299 | 102 | |
AK293780.1 | - | 23 - 24 | 2237 | 102 | |
AK303415.1 | - | 24 - 25 | 2465 | 102 | |
AK304807.1 | - | 24 - 25 | 2483 | 102 | |
AK315733.1 | - | 13 - 14 | 1234 | 102 | |
AL122037.1 | - | 1 - 2 | 72 | 102 | |
BC008844.2 | - | 7 - 8 | 671 | 102 | |
BC009355.2 | - | 14 - 15 | 1324 | 102 | |
BC063427.1 | - | 12 - 13 | 1099 | 102 | |
BC073838.1 | - | 12 - 13 | 1094 | 102 | |
BC112054.1 | - | 23 - 24 | 2338 | 102 | |
BC113605.1 | - | 23 - 24 | 2338 | 102 | |
BC143534.1 | - | 22 - 23 | 2266 | 102 | |
BC143535.1 | - | 23 - 24 | 2337 | 102 | |
BC143536.1 | - | 23 - 24 | 2331 | 102 |