Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNA1E
Entrez Gene ID
777
Gene Name
calcium voltage-gated channel subunit alpha1 E
Gene Aliases
BII, CACH6, CACNL1A6, Cav2.3, gm139
Chromosome Location
Chr.1: 181317712 - 181808084 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000721.3 NP_000712.2 22 - 23 3613 83
NM_001205293.1 NP_001192222.1 22 - 23 3613 83
NM_001205294.1 NP_001192223.1 21 - 22 3556 83
XM_017002243.1 XP_016857732.1 24 - 25 4278 83
XM_017002244.1 XP_016857733.1 24 - 25 4279 83
XM_017002245.1 XP_016857734.1 24 - 25 4279 83
XM_017002246.1 XP_016857735.1 24 - 25 4279 83
XM_017002247.1 XP_016857736.1 23 - 24 4221 83
XM_017002248.1 XP_016857737.1 24 - 25 4279 83
XM_017002249.1 XP_016857738.1 24 - 25 4278 83
XM_017002250.1 XP_016857739.1 23 - 24 4221 83
XM_017002251.1 XP_016857740.1 24 - 25 3936 83
GenBank mRNA AB209499.1 - 14 - 15 2215 83
L27745.1 - 22 - 23 3415 83
L29384.1 - 21 - 22 3526 83
L29385.1 - 22 - 23 3583 83

View Full Product Details