The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000721.3 | NP_000712.2 | 32 - 33 | 4800 | 123 |
NM_001205293.1 | NP_001192222.1 | 32 - 33 | 4800 | 123 | |
NM_001205294.1 | NP_001192223.1 | 31 - 32 | 4743 | 123 | |
XM_017002243.1 | XP_016857732.1 | 34 - 35 | 5465 | 123 | |
XM_017002244.1 | XP_016857733.1 | 34 - 35 | 5466 | 123 | |
XM_017002245.1 | XP_016857734.1 | 33 - 34 | 5454 | 123 | |
XM_017002246.1 | XP_016857735.1 | 33 - 34 | 5454 | 123 | |
XM_017002247.1 | XP_016857736.1 | 33 - 34 | 5408 | 123 | |
XM_017002248.1 | XP_016857737.1 | 34 - 35 | 5466 | 123 | |
XM_017002249.1 | XP_016857738.1 | 34 - 35 | 5465 | 123 | |
XM_017002250.1 | XP_016857739.1 | 33 - 34 | 5408 | 123 | |
XM_017002251.1 | XP_016857740.1 | 34 - 35 | 5123 | 123 | |
GenBank mRNA | AB209499.1 | - | 24 - 25 | 3402 | 123 |
L27745.1 | - | 32 - 33 | 4602 | 123 | |
L29384.1 | - | 31 - 32 | 4713 | 123 | |
L29385.1 | - | 32 - 33 | 4770 | 123 |