The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000721.3 | NP_000712.2 | 45 - 46 | 6275 | 53 |
NM_001205293.1 | NP_001192222.1 | 46 - 47 | 6404 | 53 | |
NM_001205294.1 | NP_001192223.1 | 44 - 45 | 6218 | 53 | |
XM_017002243.1 | XP_016857732.1 | 48 - 49 | 7069 | 53 | |
XM_017002244.1 | XP_016857733.1 | 48 - 49 | 7070 | 53 | |
XM_017002245.1 | XP_016857734.1 | 47 - 48 | 7058 | 53 | |
XM_017002246.1 | XP_016857735.1 | 47 - 48 | 7058 | 53 | |
XM_017002247.1 | XP_016857736.1 | 47 - 48 | 7012 | 53 | |
XM_017002248.1 | XP_016857737.1 | 47 - 48 | 7004 | 53 | |
XM_017002249.1 | XP_016857738.1 | 47 - 48 | 6940 | 53 | |
XM_017002250.1 | XP_016857739.1 | 46 - 47 | 6883 | 53 | |
XM_017002251.1 | XP_016857740.1 | 48 - 49 | 6727 | 53 | |
GenBank mRNA | AB209499.1 | - | 37 - 38 | 4877 | 53 |
L27745.1 | - | 46 - 47 | 6206 | 53 |