The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135170.1 | NP_001128642.1 | 1 - 2 | 276 | 113 |
XM_011513772.1 | XP_011512074.1 | 2 - 3 | 146 | 113 | |
GenBank mRNA | AK314802.1 | - | 1 - 2 | 224 | 113 |
BC022187.2 | - | 1 - 2 | 224 | 113 | |
BC024015.2 | - | 1 - 2 | 224 | 113 | |
BX647781.1 | - | 1 - 2 | 276 | 113 |