The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001063.3 | NP_001054.1 | 6 - 7 | 1004 | 64 |
XM_017007089.1 | XP_016862578.1 | 12 - 13 | 2354 | 64 | |
XM_017007090.1 | XP_016862579.1 | 12 - 13 | 3347 | 64 | |
GenBank mRNA | AK222755.1 | - | 6 - 7 | 744 | 64 |
AK295334.1 | - | 6 - 7 | 661 | 64 | |
AK295419.1 | - | 5 - 6 | 571 | 64 | |
AK303753.1 | - | 6 - 7 | 684 | 64 | |
BC020671.1 | - | 3 - 4 | 329 | 64 | |
BC059367.1 | - | 6 - 7 | 742 | 64 | |
BX648533.1 | - | 6 - 7 | 521 | 64 | |
CR936810.1 | - | 11 - 12 | 2425 | 64 | |
DQ923758.1 | - | 6 - 7 | 735 | 64 | |
GQ472199.1 | - | 6 - 7 | 742 | 64 | |
M12530.1 | - | 6 - 7 | 726 | 64 | |
S95936.1 | - | 6 - 7 | 775 | 64 |