Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMBT1
Entrez Gene ID
1755
Gene Name
deleted in malignant brain tumors 1
Gene Aliases
GP340, SAG, muclin
Chromosome Location
Chr.10: 122560665 - 122643736 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320644.1 NP_001307573.1 10 - 11 1110 79
NM_004406.2 NP_004397.2 10 - 11 1110 79
NM_007329.2 NP_015568.2 10 - 11 1110 79
NM_017579.2 NP_060049.2 10 - 11 1110 79
XM_006717660.3 XP_006717723.1 9 - 10 1050 79
XM_006717665.3 XP_006717728.1 8 - 9 1002 79
XM_011539388.2 XP_011537690.1 10 - 11 1098 79
XM_011539389.2 XP_011537691.1 10 - 11 1098 79
XM_011539390.2 XP_011537692.1 10 - 11 1098 79
XM_011539391.2 XP_011537693.1 10 - 11 1098 79
XM_011539392.2 XP_011537694.1 10 - 11 1098 79
XM_011539393.2 XP_011537695.1 10 - 11 1098 79
XM_011539394.2 XP_011537696.1 10 - 11 1098 79
XM_011539395.2 XP_011537697.1 10 - 11 1098 79
XM_011539396.2 XP_011537698.1 9 - 10 1050 79
XM_011539399.2 XP_011537701.1 8 - 9 1002 79
XM_011539400.2 XP_011537702.1 10 - 11 1098 79
XM_011539401.2 XP_011537703.1 10 - 11 1098 79
XM_011539403.2 XP_011537705.1 10 - 11 1098 79
XM_011539405.2 XP_011537707.1 10 - 11 1099 79
XM_011539407.2 XP_011537709.1 10 - 11 1098 79
XM_011539408.2 XP_011537710.1 10 - 11 1098 79
XM_011539409.2 XP_011537711.1 10 - 11 1098 79
XM_011539410.2 XP_011537712.1 10 - 11 1098 79
XM_011539411.2 XP_011537713.1 10 - 11 1098 79
XM_011539413.2 XP_011537715.1 10 - 11 1097 79
XM_011539415.2 XP_011537717.1 10 - 11 1100 79
GenBank mRNA AB209691.1 - 8 - 9 900 79
AF159456.1 - 10 - 11 1110 79
AJ000342.1 - 10 - 11 1110 79
AJ243212.1 - 10 - 11 1110 79
AJ243224.1 - 10 - 11 79
AJ297935.1 - 10 - 11 1091 79
BC153299.1 - 10 - 11 1097 79

View Full Product Details