Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DMBT1
Entrez Gene ID
1755
Gene Name
deleted in malignant brain tumors 1
Gene Aliases
GP340, SAG, muclin
Chromosome Location
Chr.10: 122560665 - 122643736 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320644.1 NP_001307573.1 44 - 45 5541 152
NM_004406.2 NP_004397.2 31 - 32 3657 152
NM_007329.2 NP_015568.2 44 - 45 5541 152
NM_017579.2 NP_060049.2 43 - 44 5511 152
XM_006717660.3 XP_006717723.1 46 - 47 5868 152
XM_006717665.3 XP_006717728.1 45 - 46 5820 152
XM_011539388.2 XP_011537690.1 47 - 48 5916 152
XM_011539389.2 XP_011537691.1 47 - 48 5916 152
XM_011539390.2 XP_011537692.1 46 - 47 5886 152
XM_011539391.2 XP_011537693.1 46 - 47 5886 152
XM_011539392.2 XP_011537694.1 46 - 47 5886 152
XM_011539393.2 XP_011537695.1 46 - 47 5886 152
XM_011539394.2 XP_011537696.1 46 - 47 5886 152
XM_011539395.2 XP_011537697.1 46 - 47 5886 152
XM_011539396.2 XP_011537698.1 46 - 47 5868 152
XM_011539398.2 XP_011537700.1 45 - 46 5853 152
XM_011539399.2 XP_011537701.1 45 - 46 5820 152
XM_011539400.2 XP_011537702.1 46 - 47 5592 152
XM_011539401.2 XP_011537703.1 44 - 45 5529 152
XM_011539402.2 XP_011537704.1 44 - 45 5529 152
XM_011539403.2 XP_011537705.1 44 - 45 5529 152
XM_011539405.2 XP_011537707.1 44 - 45 5530 152
XM_011539407.2 XP_011537709.1 44 - 45 5523 152
XM_011539408.2 XP_011537710.1 44 - 45 5523 152
XM_011539409.2 XP_011537711.1 44 - 45 5523 152
XM_011539410.2 XP_011537712.1 44 - 45 5523 152
XM_011539411.2 XP_011537713.1 43 - 44 5499 152
XM_011539413.2 XP_011537715.1 41 - 42 5135 152
XM_011539414.2 XP_011537716.1 31 - 32 3636 152
XM_011539415.2 XP_011537717.1 29 - 30 3590 152
XM_017015798.1 XP_016871287.1 32 - 33 4178 152
GenBank mRNA AF159456.1 - 44 - 45 5541 152
AJ000342.1 - 31 - 32 3657 152
AJ243212.1 - 44 - 45 5541 152
AJ243224.1 - 43 - 44 5511 152
AJ297935.1 - 44 - 45 5522 152
AK304149.1 - 18 - 19 1681 152
BC153299.1 - 28 - 29 3275 152
BX640988.1 - 2 - 3 313 152

View Full Product Details