Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMBT1
Entrez Gene ID
1755
Gene Name
deleted in malignant brain tumors 1
Gene Aliases
GP340, SAG, muclin
Chromosome Location
Chr.10: 122560665 - 122643736 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320644.1 NP_001307573.1 35 - 36 4550 78
NM_004406.2 NP_004397.2 22 - 23 2666 78
NM_007329.2 NP_015568.2 35 - 36 4550 78
NM_017579.2 NP_060049.2 34 - 35 4520 78
XM_006717660.3 XP_006717723.1 37 - 38 4877 78
XM_006717665.3 XP_006717728.1 36 - 37 4829 78
XM_011539388.2 XP_011537690.1 38 - 39 4925 78
XM_011539389.2 XP_011537691.1 38 - 39 4925 78
XM_011539390.2 XP_011537692.1 37 - 38 4895 78
XM_011539391.2 XP_011537693.1 37 - 38 4895 78
XM_011539392.2 XP_011537694.1 37 - 38 4895 78
XM_011539393.2 XP_011537695.1 37 - 38 4895 78
XM_011539394.2 XP_011537696.1 37 - 38 4895 78
XM_011539395.2 XP_011537697.1 37 - 38 4895 78
XM_011539396.2 XP_011537698.1 37 - 38 4877 78
XM_011539398.2 XP_011537700.1 36 - 37 4862 78
XM_011539399.2 XP_011537701.1 36 - 37 4829 78
XM_011539400.2 XP_011537702.1 37 - 38 4601 78
XM_011539401.2 XP_011537703.1 35 - 36 4538 78
XM_011539402.2 XP_011537704.1 35 - 36 4538 78
XM_011539405.2 XP_011537707.1 35 - 36 4539 78
XM_011539407.2 XP_011537709.1 35 - 36 4532 78
XM_011539408.2 XP_011537710.1 35 - 36 4532 78
XM_011539409.2 XP_011537711.1 35 - 36 4532 78
XM_011539410.2 XP_011537712.1 38 - 39 4925 78
XM_011539411.2 XP_011537713.1 34 - 35 4508 78
XM_011539413.2 XP_011537715.1 32 - 33 4144 78
XM_011539414.2 XP_011537716.1 22 - 23 2645 78
XM_017015798.1 XP_016871287.1 23 - 24 3187 78
GenBank mRNA AF159456.1 - 35 - 36 4550 78
AJ000342.1 - 22 - 23 2666 78
AJ243224.1 - 34 - 35 4520 78
AJ297935.1 - 35 - 36 4531 78
AK304149.1 - 11 - 12 1083 78
BC153299.1 - 19 - 20 2284 78

View Full Product Details