The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001436.3 | NP_001427.2 | 5 - 6 | 662 | 65 |
XM_005258651.2 | XP_005258708.1 | 5 - 6 | 640 | 65 | |
XM_011526623.2 | XP_011524925.1 | 4 - 5 | 443 | 65 | |
GenBank mRNA | AB451384.1 | - | 4 - 5 | 548 | 65 |
AK296950.1 | - | 5 - 6 | 627 | 65 | |
BC015218.2 | - | 3 - 4 | 367 | 65 | |
BC019260.1 | - | 5 - 6 | 616 | 65 | |
BC019609.1 | - | 3 - 4 | 366 | 65 | |
BT006830.1 | - | 4 - 5 | 548 | 65 | |
BT020144.1 | - | 4 - 5 | 548 | 65 | |
CR457069.1 | - | 4 - 5 | 548 | 65 | |
M59849.1 | - | 5 - 6 | 576 | 65 | |
X56597.1 | - | 5 - 6 | 607 | 65 |