Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBL
Entrez Gene ID
2091
Gene Name
fibrillarin
Gene Aliases
FIB, FLRN, RNU3IP1
Chromosome Location
Chr.19: 39834453 - 39846414 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001436.3 NP_001427.2 7 - 8 911 63
XM_005258651.2 XP_005258708.1 7 - 8 889 63
XM_011526623.2 XP_011524925.1 6 - 7 692 63
GenBank mRNA AB451384.1 - 6 - 7 797 63
AK296950.1 - 7 - 8 876 63
BC015218.2 - 5 - 6 616 63
BC019260.1 - 7 - 8 865 63
BC019609.1 - 5 - 6 615 63
BT006830.1 - 6 - 7 797 63
BT020144.1 - 6 - 7 797 63
CR457069.1 - 6 - 7 797 63
M59849.1 - 7 - 8 825 63
X56597.1 - 7 - 8 856 63

View Full Product Details