Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM27
Entrez Gene ID
57393
Gene Name
transmembrane protein 27
Gene Aliases
NX-17, NX17
Chromosome Location
Chr.X: 15627316 - 15675624 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020665.5 NP_065716.1 4 - 5 566 66
XM_017029680.1 XP_016885169.1 4 - 5 434 66
GenBank mRNA AK313835.1 - 4 - 5 391 66
AY359060.1 - 4 - 5 316 66
BC014317.1 - 4 - 5 570 66
BC015099.1 - 4 - 5 398 66
BC050606.1 - 4 - 5 318 66

View Full Product Details