The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105513.2 | NP_001098983.2 | 23 - 24 | 3305 | 65 |
NM_145755.2 | NP_665698.2 | 24 - 25 | 3449 | 65 | |
XM_005264922.4 | XP_005264979.1 | 24 - 25 | 3405 | 65 | |
XM_005264923.3 | XP_005264980.1 | 24 - 25 | 3378 | 65 | |
XM_017005843.1 | XP_016861332.1 | 21 - 22 | 3024 | 65 | |
GenBank mRNA | AK093313.1 | - | 14 - 15 | 1989 | 65 |
AK098528.1 | - | 6 - 7 | 815 | 65 | |
AK302408.1 | - | 23 - 24 | 3288 | 65 | |
AK302465.1 | - | 23 - 24 | 3159 | 65 | |
BC129948.1 | - | 24 - 25 | 3360 | 65 | |
BX647142.1 | - | 5 - 6 | 1021 | 65 |