The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_058165.2 | NP_477513.2 | 4 - 5 | 705 | 110 |
GenBank mRNA | AF384163.1 | - | 4 - 5 | 702 | 110 |