The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321643.1 | NP_001308572.1 | 32 - 33 | 4153 | 90 |
NM_004850.4 | NP_004841.2 | 32 - 33 | 4609 | 90 | |
GenBank mRNA | AB014519.1 | - | 32 - 33 | 4614 | 90 |
AK299296.1 | - | 18 - 19 | 2616 | 90 | |
AK310358.1 | - | 28 - 29 | 3904 | 90 | |
BC111801.1 | - | 19 - 20 | 2716 | 90 | |
D87931.1 | - | 32 - 33 | 4160 | 90 |