The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016052.3 | NP_057136.2 | 4 - 5 | 738 | 93 |
GenBank mRNA | AF151873.1 | - | 4 - 5 | 673 | 93 |
AK290066.1 | - | 4 - 5 | 720 | 93 | |
BC020641.1 | - | 4 - 5 | 715 | 93 |