Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INPPL1
Entrez Gene ID
3636
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
Chromosome Location
Chr.11: 72223861 - 72239147 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001567.3 NP_001558.3 27 - 28 3830 77
XM_005273978.3 XP_005274035.1 28 - 29 4207 77
XM_005273979.4 XP_005274036.1 28 - 29 4054 77
XM_011544999.2 XP_011543301.1 28 - 29 3988 77
XM_017017673.1 XP_016873162.1 29 - 30 4145 77
GenBank mRNA BC140853.1 - 27 - 28 3830 77
L24444.1 - 2 - 3 420 77
L36818.1 - 26 - 27 3808 77
Y14385.1 - 27 - 28 3894 77

View Full Product Details