The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001567.3 | NP_001558.3 | 2 - 3 | 394 | 106 |
XM_005273978.3 | XP_005274035.1 | 3 - 4 | 705 | 106 | |
XM_005273979.4 | XP_005274036.1 | 3 - 4 | 552 | 106 | |
XM_011544999.2 | XP_011543301.1 | 3 - 4 | 552 | 106 | |
XM_011545000.2 | XP_011543302.1 | 3 - 4 | 552 | 106 | |
XM_017017673.1 | XP_016873162.1 | 4 - 5 | 643 | 106 | |
GenBank mRNA | BC140853.1 | - | 2 - 3 | 394 | 106 |
L36818.1 | - | 1 - 2 | 370 | 106 | |
Y14385.1 | - | 2 - 3 | 458 | 106 |