Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRC5
Entrez Gene ID
84166
Gene Name
NLR family CARD domain containing 5
Gene Aliases
CLR16.1, NOD27, NOD4
Chromosome Location
Chr.16: 56989547 - 57083524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032206.4 NP_115582.4 9 - 10 2543 97
XM_005256193.2 XP_005256250.1 10 - 11 2604 97
XM_005256194.3 XP_005256251.1 10 - 11 2815 97
XM_005256195.2 XP_005256252.1 10 - 11 2604 97
XM_005256196.2 XP_005256253.1 9 - 10 2520 97
XM_005256197.2 XP_005256254.1 10 - 11 2604 97
XM_005256199.3 XP_005256256.1 10 - 11 2604 97
XM_005256201.3 XP_005256258.1 10 - 11 2604 97
XM_006721297.2 XP_006721360.1 10 - 11 2592 97
XM_006721298.2 XP_006721361.1 10 - 11 2604 97
XM_006721300.2 XP_006721363.1 10 - 11 2604 97
XM_011523373.2 XP_011521675.1 10 - 11 2751 97
XM_011523375.2 XP_011521677.1 10 - 11 2604 97
XM_011523376.2 XP_011521678.1 5 - 6 430 97
XM_017023766.1 XP_016879255.1 10 - 11 2604 97
XM_017023767.1 XP_016879256.1 10 - 11 2592 97
XM_017023768.1 XP_016879257.1 10 - 11 2604 97
XM_017023769.1 XP_016879258.1 10 - 11 2592 97
XM_017023770.1 XP_016879259.1 10 - 11 2604 97
XM_017023771.1 XP_016879260.1 10 - 11 2604 97
GenBank mRNA AK025362.1 - 5 - 6 1675 97
AK090439.1 - 5 - 6 1598 97
BX640842.1 - 6 - 7 1078 97

View Full Product Details