Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRC5
Entrez Gene ID
84166
Gene Name
NLR family CARD domain containing 5
Gene Aliases
CLR16.1, NOD27, NOD4
Chromosome Location
Chr.16: 56989547 - 57083524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032206.4 NP_115582.4 22 - 23 3550 73
XM_005256193.2 XP_005256250.1 23 - 24 3611 73
XM_005256194.3 XP_005256251.1 23 - 24 3822 73
XM_005256195.2 XP_005256252.1 23 - 24 3611 73
XM_005256196.2 XP_005256253.1 22 - 23 3527 73
XM_005256197.2 XP_005256254.1 23 - 24 3611 73
XM_005256199.3 XP_005256256.1 23 - 24 3611 73
XM_005256201.3 XP_005256258.1 22 - 23 3521 73
XM_006721297.2 XP_006721360.1 23 - 24 3599 73
XM_006721298.2 XP_006721361.1 22 - 23 3521 73
XM_006721300.2 XP_006721363.1 23 - 24 3611 73
XM_011523373.2 XP_011521675.1 23 - 24 3758 73
XM_011523375.2 XP_011521677.1 23 - 24 3611 73
XM_011523376.2 XP_011521678.1 18 - 19 1437 73
XM_017023766.1 XP_016879255.1 23 - 24 3611 73
XM_017023767.1 XP_016879256.1 23 - 24 3599 73
XM_017023768.1 XP_016879257.1 23 - 24 3611 73
XM_017023769.1 XP_016879258.1 23 - 24 3599 73
XM_017023770.1 XP_016879259.1 23 - 24 3611 73
XM_017023771.1 XP_016879260.1 23 - 24 3611 73
GenBank mRNA AF389420.1 - 21 - 22 3423 73
AK025362.1 - 18 - 19 2682 73
AK027414.1 - 3 - 4 227 73
AK027416.1 - 3 - 4 226 73
AK074133.1 - 11 - 12 876 73
AK090439.1 - 18 - 19 2605 73
BC063566.1 - 5 - 6 336 73
EF452236.1 - 21 - 22 3423 73

View Full Product Details