The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021094.3 | NP_066580.1 | 9 - 10 | 1517 | 165 |
NM_134431.3 | NP_602307.1 | 11 - 12 | 1987 | 165 | |
XM_005253474.3 | XP_005253531.1 | 10 - 11 | 1408 | 165 | |
XM_005253477.3 | XP_005253534.1 | 9 - 10 | 1295 | 165 | |
XM_011520818.1 | XP_011519120.1 | 10 - 11 | 1415 | 165 | |
XM_011520819.1 | XP_011519121.1 | 10 - 11 | 1562 | 165 | |
XM_011520820.2 | XP_011519122.1 | 10 - 11 | 2884 | 165 | |
XM_011520821.2 | XP_011519123.1 | 3 - 4 | 1687 | 165 | |
XM_017019849.1 | XP_016875338.1 | 10 - 11 | 1415 | 165 | |
XM_017019850.1 | XP_016875339.1 | 9 - 10 | 1295 | 165 | |
GenBank mRNA | AF085224.1 | - | 10 - 11 | 1476 | 165 |
AF279784.1 | - | 11 - 12 | 1987 | 165 | |
AK296044.1 | - | 8 - 9 | 1128 | 165 | |
AK316249.1 | - | 9 - 10 | 1547 | 165 | |
U21943.1 | - | 9 - 10 | 1319 | 165 |