The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025237.2 | NP_079513.1 | 1 - 2 | 272 | 51 |
GenBank mRNA | AF326739.1 | - | 1 - 2 | 272 | 51 |
AF331844.1 | - | 1 - 2 | 262 | 51 | |
AY358627.1 | - | 1 - 2 | 286 | 51 | |
BC101086.1 | - | 1 - 2 | 252 | 51 | |
BC101087.1 | - | 1 - 2 | 252 | 51 | |
BC101088.1 | - | 1 - 2 | 252 | 51 | |
BC101089.1 | - | 1 - 2 | 252 | 51 |