Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXP2
Entrez Gene ID
93986
Gene Name
forkhead box P2
Gene Aliases
CAGH44, SPCH1, TNRC10
Chromosome Location
Chr.7: 114086310 - 114693772 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172766.2 NP_001166237.1 2 - 3 542 78
NM_001172767.2 NP_001166238.1 1 - 2 178 78
NM_014491.3 NP_055306.1 2 - 3 542 78
NM_148898.3 NP_683696.2 2 - 3 542 78
NM_148899.3 NP_683697.2 1 - 2 178 78
NM_148900.3 NP_683698.2 2 - 3 542 78
NR_033766.1 3 - 4 571 78
NR_033767.1 2 - 3 542 78
XM_017012801.1 XP_016868290.1 4 - 5 1148 78
GenBank mRNA AF337817.1 - 2 - 3 318 78
AF467252.1 - 2 - 3 269 78
AF467253.1 - 1 - 2 180 78
AF467257.1 - 2 - 3 220 78
AF467258.1 - 1 - 2 165 78
AF493430.1 - 2 - 3 335 78
AK131266.1 - 3 - 4 555 78
AK296957.1 - 2 - 3 422 78
BC126104.1 - 2 - 3 216 78
BC143866.1 - 2 - 3 216 78
BC143867.1 - 2 - 3 216 78
CR749236.1 - 6 - 7 662 78
KJ534839.1 - 1 - 2 168 78
KJ535079.1 - 1 - 2 168 78
KJ535086.1 - 1 - 2 167 78

View Full Product Details