The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172766.2 | NP_001166237.1 | 9 - 10 | 1560 | 72 |
NM_001172767.2 | NP_001166238.1 | 9 - 10 | 1274 | 72 | |
NM_014491.3 | NP_055306.1 | 9 - 10 | 1563 | 72 | |
NM_148898.3 | NP_683696.2 | 10 - 11 | 1638 | 72 | |
NM_148899.3 | NP_683697.2 | 8 - 9 | 1199 | 72 | |
NM_148900.3 | NP_683698.2 | 10 - 11 | 1614 | 72 | |
NR_033766.1 | 13 - 14 | 1776 | 72 | ||
NR_033767.1 | 10 - 11 | 1621 | 72 | ||
XM_017012801.1 | XP_016868290.1 | 12 - 13 | 2244 | 72 | |
GenBank mRNA | AF337817.1 | - | 9 - 10 | 1339 | 72 |
AF467253.1 | - | 8 - 9 | 1201 | 72 | |
AF467254.1 | - | 2 - 3 | 180 | 72 | |
AF467255.1 | - | 3 - 4 | 416 | 72 | |
AF467256.1 | - | 2 - 3 | 180 | 72 | |
AF467258.1 | - | 8 - 9 | 1186 | 72 | |
AF467259.1 | - | 9 - 10 | 1261 | 72 | |
AK131266.1 | - | 13 - 14 | 1760 | 72 | |
AK296957.1 | - | 10 - 11 | 1494 | 72 | |
AY144615.1 | - | 10 - 11 | 1633 | 72 | |
BC126104.1 | - | 10 - 11 | 1295 | 72 | |
BC143866.1 | - | 10 - 11 | 1295 | 72 | |
BC143867.1 | - | 9 - 10 | 1234 | 72 | |
CR749236.1 | - | 15 - 16 | 1789 | 72 | |
DQ778626.1 | - | 6 - 7 | 913 | 72 | |
KJ534839.1 | - | 9 - 10 | 1264 | 72 | |
KJ535079.1 | - | 8 - 9 | 1402 | 72 | |
KJ535086.1 | - | 8 - 9 | 1188 | 72 |