The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261406.1 | NP_001248335.1 | 32 - 33 | 4679 | 113 |
NM_001261407.1 | NP_001248336.1 | 32 - 33 | 4679 | 113 | |
NM_003829.4 | NP_003820.2 | 33 - 34 | 4778 | 113 | |
XM_005251622.4 | XP_005251679.1 | 33 - 34 | 4727 | 113 | |
XM_005251623.4 | XP_005251680.1 | 31 - 32 | 4517 | 113 | |
XM_006716885.3 | XP_006716948.1 | 33 - 34 | 4776 | 113 | |
XM_006716886.3 | XP_006716949.1 | 34 - 35 | 4935 | 113 | |
XM_006716887.3 | XP_006716950.1 | 33 - 34 | 4879 | 113 | |
XM_006716888.3 | XP_006716951.1 | 33 - 34 | 4727 | 113 | |
XM_006716889.3 | XP_006716952.1 | 32 - 33 | 4631 | 113 | |
XM_006716891.3 | XP_006716954.1 | 32 - 33 | 4631 | 113 | |
XM_017015252.1 | XP_016870741.1 | 33 - 34 | 4727 | 113 | |
XM_017015253.1 | XP_016870742.1 | 32 - 33 | 4616 | 113 | |
XM_017015254.1 | XP_016870743.1 | 32 - 33 | 4728 | 113 | |
XM_017015255.1 | XP_016870744.1 | 31 - 32 | 4576 | 113 | |
XM_017015256.1 | XP_016870745.1 | 31 - 32 | 4517 | 113 | |
XM_017015257.1 | XP_016870746.1 | 31 - 32 | 4504 | 113 | |
GenBank mRNA | AB210041.1 | - | 33 - 34 | 4779 | 113 |
AF093419.1 | - | 32 - 33 | 4602 | 113 | |
AK074721.1 | - | 3 - 4 | 263 | 113 | |
AK091945.1 | - | 8 - 9 | 1056 | 113 | |
AK294823.1 | - | 6 - 7 | 688 | 113 | |
AK294976.1 | - | 11 - 12 | 1284 | 113 | |
AK307988.1 | - | 9 - 10 | 1096 | 113 | |
AK316489.1 | - | 10 - 11 | 1162 | 113 | |
BC140793.1 | - | 31 - 32 | 4503 | 113 | |
BC144564.1 | - | 31 - 32 | 4503 | 113 | |
CR936648.1 | - | 17 - 18 | 4666 | 113 |