Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPDZ
Entrez Gene ID
8777
Gene Name
multiple PDZ domain crumbs cell polarity complex component
Gene Aliases
HYC2, MUPP1
Chromosome Location
Chr.9: 13105296 - 13279687 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261406.1 NP_001248335.1 45 - 46 6194 97
NM_001261407.1 NP_001248336.1 44 - 45 6107 97
NM_003829.4 NP_003820.2 45 - 46 6206 97
XM_005251622.4 XP_005251679.1 46 - 47 6242 97
XM_005251623.4 XP_005251680.1 44 - 45 6032 97
XM_006716885.3 XP_006716948.1 46 - 47 6291 97
XM_006716886.3 XP_006716949.1 47 - 48 6450 97
XM_006716887.3 XP_006716950.1 46 - 47 6394 97
XM_006716888.3 XP_006716951.1 45 - 46 6155 97
XM_006716889.3 XP_006716952.1 45 - 46 6146 97
XM_006716891.3 XP_006716954.1 44 - 45 6059 97
XM_017015252.1 XP_016870741.1 45 - 46 6158 97
XM_017015253.1 XP_016870742.1 44 - 45 6044 97
XM_017015254.1 XP_016870743.1 45 - 46 6243 97
XM_017015255.1 XP_016870744.1 44 - 45 6091 97
XM_017015256.1 XP_016870745.1 43 - 44 5945 97
XM_017015257.1 XP_016870746.1 43 - 44 5932 97
GenBank mRNA AB210041.1 - 45 - 46 6207 97
AF093419.1 - 45 - 46 6033 97
AJ001319.1 - 11 - 12 1220 97
AK058011.1 - 6 - 7 1633 97
AK074721.1 - 15 - 16 1695 97
AK091945.1 - 21 - 22 2571 97
AK098775.1 - 13 - 14 1471 97
AK294823.1 - 19 - 20 2203 97
AK294976.1 - 24 - 25 2799 97
AK307988.1 - 22 - 23 2611 97
AK316489.1 - 23 - 24 2677 97
BC140793.1 - 44 - 45 6018 97
BC144564.1 - 43 - 44 5931 97
CR936648.1 - 30 - 31 6181 97
CX782625.1 - 1 - 2 54 97

View Full Product Details