Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NALCN
Entrez Gene ID
259232
Gene Name
sodium leak channel, non-selective
Gene Aliases
CLIFAHDD, CanIon, IHPRF, IHPRF1, INNFD, VGCNL1, bA430M15.1
Chromosome Location
Chr.13: 101053774 - 101417206 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052867.2 NP_443099.1 11 - 12 1460 78
XM_011521067.2 XP_011519369.1 11 - 12 1400 78
XM_011521068.2 XP_011519370.1 11 - 12 1383 78
XM_011521069.2 XP_011519371.1 10 - 11 1313 78
XM_017020534.1 XP_016876023.1 10 - 11 1344 78
XM_017020535.1 XP_016876024.1 10 - 11 1286 78
XM_017020536.1 XP_016876025.1 6 - 7 1212 78
XM_017020537.1 XP_016876026.1 5 - 6 601 78
GenBank mRNA AK094390.1 - 2 - 3 220 78
AK094669.1 - 11 - 12 1431 78
AY141972.1 - 11 - 12 1354 78
BC028390.1 - 11 - 12 1367 78

View Full Product Details