The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052867.2 | NP_443099.1 | 38 - 39 | 4520 | 86 |
XM_011521067.2 | XP_011519369.1 | 38 - 39 | 4460 | 86 | |
XM_011521068.2 | XP_011519370.1 | 38 - 39 | 4443 | 86 | |
XM_011521069.2 | XP_011519371.1 | 37 - 38 | 4373 | 86 | |
XM_017020534.1 | XP_016876023.1 | 37 - 38 | 4404 | 86 | |
XM_017020535.1 | XP_016876024.1 | 37 - 38 | 4346 | 86 | |
XM_017020536.1 | XP_016876025.1 | 33 - 34 | 4272 | 86 | |
XM_017020537.1 | XP_016876026.1 | 32 - 33 | 3661 | 86 | |
GenBank mRNA | AK126621.1 | - | 19 - 20 | 1984 | 86 |
AK126718.1 | - | 11 - 12 | 1121 | 86 | |
AK172752.1 | - | 1 - 2 | 51 | 86 | |
AY141972.1 | - | 38 - 39 | 4414 | 86 |