The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322194.1 | NP_001309123.1 | 20 - 21 | 3356 | 166 |
NM_001322195.1 | NP_001309124.1 | 19 - 20 | 2874 | 166 | |
NM_001322196.1 | NP_001309125.1 | 19 - 20 | 3187 | 166 | |
NM_001322198.1 | NP_001309127.1 | 20 - 21 | 3175 | 166 | |
NM_001322199.1 | NP_001309128.1 | 20 - 21 | 3117 | 166 | |
NM_001322204.1 | NP_001309133.1 | 17 - 18 | 2626 | 166 | |
NM_004972.3 | NP_004963.1 | 20 - 21 | 3255 | 166 | |
GenBank mRNA | AF001362.1 | - | 18 - 19 | 2761 | 166 |
AF005216.1 | - | 20 - 21 | 3255 | 166 | |
AF058925.1 | - | 20 - 21 | 3238 | 166 | |
AK292525.1 | - | 19 - 20 | 2874 | 166 | |
AK302618.1 | - | 17 - 18 | 2685 | 166 | |
AY973034.1 | - | 19 - 20 | 2832 | 166 | |
BC039695.1 | - | 19 - 20 | 2859 | 166 |