The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017582.6 | NP_060052.3 | 11 - 12 | 1256 | 119 |
GenBank mRNA | AF116721.1 | - | 4 - 5 | 232 | 119 |
AJ243666.1 | - | 5 - 6 | 323 | 119 | |
AK056388.1 | - | 7 - 8 | 556 | 119 | |
AK293986.1 | - | 11 - 12 | 879 | 119 | |
AY112698.1 | - | 11 - 12 | 1231 | 119 | |
AY948200.1 | - | 11 - 12 | 1239 | 119 | |
BC000848.1 | - | 7 - 8 | 590 | 119 | |
BC009286.2 | - | 6 - 7 | 394 | 119 | |
BC015316.1 | - | 3 - 4 | 199 | 119 | |
BC061583.1 | - | 7 - 8 | 568 | 119 | |
BC070158.1 | - | 10 - 11 | 781 | 119 | |
BC087836.1 | - | 9 - 10 | 667 | 119 | |
BC107057.2 | - | 12 - 13 | 924 | 119 | |
BC148185.2 | - | 11 - 12 | 933 | 119 | |
CR457219.1 | - | 2 - 3 | 112 | 119 |