Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HAPLN2
Entrez Gene ID
60484
Gene Name
hyaluronan and proteoglycan link protein 2
Gene Aliases
BRAL1
Chromosome Location
Chr.1: 156601504 - 156626608 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005245415.2 XP_005245472.1 1 - 2 165 73
XM_017002021.1 XP_016857510.1 1 - 2 165 73
GenBank mRNA AK293797.1 - 1 - 2 85 73
AK308980.1 - 1 - 2 106 73

View Full Product Details