The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001218.4 | NP_001209.1 | 4 - 5 | 818 | 73 |
NM_001293642.1 | NP_001280571.1 | 3 - 4 | 638 | 73 | |
NM_206925.2 | NP_996808.1 | 4 - 5 | 818 | 73 | |
NR_135511.1 | 4 - 5 | 921 | 73 | ||
GenBank mRNA | AF037335.1 | - | 4 - 5 | 543 | 73 |
AF051882.1 | - | 4 - 5 | 435 | 73 | |
AK096845.1 | - | 3 - 4 | 374 | 73 | |
AK315769.1 | - | 4 - 5 | 584 | 73 | |
BC000278.1 | - | 4 - 5 | 540 | 73 | |
BC011691.2 | - | 4 - 5 | 543 | 73 | |
BC023981.1 | - | 4 - 5 | 558 | 73 | |
BT006656.1 | - | 4 - 5 | 428 | 73 |