Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CA12
Entrez Gene ID
771
Gene Name
carbonic anhydrase 12
Gene Aliases
CA-XII, CAXII, HsT18816, T18816
Chromosome Location
Chr.15: 63321378 - 63382166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293642.1 NP_001280571.1 2 - 3 498 57
GenBank mRNA AK096845.1 - 2 - 3 234 57

View Full Product Details