Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
R3HDM2
Entrez Gene ID
22864
Gene Name
R3H domain containing 2
Gene Aliases
CAG6, PR01365
Chromosome Location
Chr.12: 57253764 - 57430999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014925.3 NP_055740.3 19 - 20 2446 83
XM_005268725.3 XP_005268782.1 21 - 22 2796 83
XM_011538031.1 XP_011536333.1 22 - 23 2684 83
XM_011538033.1 XP_011536335.1 23 - 24 2738 83
XM_011538035.1 XP_011536337.1 22 - 23 2642 83
XM_011538036.1 XP_011536338.1 22 - 23 2636 83
XM_011538037.1 XP_011536339.1 21 - 22 2618 83
XM_011538038.1 XP_011536340.1 21 - 22 2588 83
XM_011538039.1 XP_011536341.1 21 - 22 2582 83
XM_011538040.1 XP_011536342.1 22 - 23 2636 83
XM_011538042.2 XP_011536344.1 24 - 25 2834 83
XM_017018998.1 XP_016874487.1 21 - 22 2660 83
XM_017018999.1 XP_016874488.1 24 - 25 2875 83
XM_017019000.1 XP_016874489.1 22 - 23 2642 83
XM_017019001.1 XP_016874490.1 23 - 24 2948 83
XM_017019002.1 XP_016874491.1 21 - 22 2612 83
XM_017019003.1 XP_016874492.1 21 - 22 2588 83
XM_017019004.1 XP_016874493.1 21 - 22 2582 83
XM_017019005.1 XP_016874494.1 23 - 24 2793 83
XM_017019006.1 XP_016874495.1 22 - 23 2686 83
XM_017019007.1 XP_016874496.1 22 - 23 2721 83
XM_017019008.1 XP_016874497.1 20 - 21 2486 83
XM_017019009.1 XP_016874498.1 21 - 22 2540 83
XM_017019010.1 XP_016874499.1 21 - 22 2614 83
XM_017019011.1 XP_016874500.1 24 - 25 2837 83
XM_017019012.1 XP_016874501.1 20 - 21 2522 83
XM_017019013.1 XP_016874502.1 21 - 22 2656 83
XM_017019014.1 XP_016874503.1 21 - 22 2691 83
XM_017019015.1 XP_016874504.1 21 - 22 2833 83
XM_017019016.1 XP_016874505.1 22 - 23 2852 83
XM_017019017.1 XP_016874506.1 21 - 22 2614 83
XM_017019018.1 XP_016874507.1 21 - 22 2580 83
XM_017019019.1 XP_016874508.1 22 - 23 2676 83
XM_017019020.1 XP_016874509.1 19 - 20 2426 83
XM_017019021.1 XP_016874510.1 21 - 22 2650 83
XM_017019022.1 XP_016874511.1 20 - 21 2560 83
XM_017019023.1 XP_016874512.1 20 - 21 2571 83
XM_017019024.1 XP_016874513.1 20 - 21 2560 83
XM_017019025.1 XP_016874514.1 20 - 21 2525 83
XM_017019026.1 XP_016874515.1 21 - 22 2565 83
XM_017019027.1 XP_016874516.1 20 - 21 2696 83
XM_017019028.1 XP_016874517.1 19 - 20 2458 83
XM_017019029.1 XP_016874518.1 19 - 20 2544 83
XM_017019030.1 XP_016874519.1 12 - 13 1825 83
XM_017019031.1 XP_016874520.1 15 - 16 2109 83
XM_017019032.1 XP_016874521.1 11 - 12 1677 83
XM_017019033.1 XP_016874522.1 13 - 14 1828 83
GenBank mRNA AB023219.1 - 21 - 22 2803 83
AK293394.1 - 11 - 12 1698 83
AX775761.1 - 21 - 22 2803 83
AX775763.1 - 21 - 22 2845 83
BC041857.1 - 11 - 12 2258 83
BC104995.1 - 9 - 10 1453 83
BC112226.1 - 9 - 10 1453 83
BC143605.1 - 10 - 11 1555 83

View Full Product Details