Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- R3HDM2
- Entrez Gene ID
-
22864
- Gene Name
- R3H domain containing 2
- Gene Aliases
-
CAG6, PR01365
- Chromosome Location
-
Chr.12: 57253764 - 57430999 on Build GRCh38
View Full Product Details