Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TMCC1
- Entrez Gene ID
-
23023
- Gene Name
- transmembrane and coiled-coil domain family 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 129647792 - 129893605 on Build GRCh38
View Full Product Details