Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMCC1
Entrez Gene ID
23023
Gene Name
transmembrane and coiled-coil domain family 1
Gene Aliases
-
Chromosome Location
Chr.3: 129647792 - 129893605 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128224.2 NP_001121696.1 2 - 3 135 100
GenBank mRNA CA389758.1 - 2 - 3 119 100

View Full Product Details